Assay Details
| Gene Symbol: HRH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018057589.1 | XP_017913078.1 | 2 - 3 | 727 | 100 | |
| GenBank mRNA | JO621326.2 | - | 2 - 3 | 403 | 100 | |
More Information
Set Membership: |
|

HRH3
Genomic Map