Assay Details
Gene Symbol: RCN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062791.1 | XP_017918280.1 | 5 - 6 | 1082 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
