Assay Details
| Gene Symbol: NSUN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005674810.3 | XP_005674867.1 | 5 - 6 | 889 | 87 | |
| GenBank mRNA | JT716494.1 | - | 6 - 7 | 704 | 87 | |
More Information
Set Membership: |
|

NSUN3
Genomic Map