Assay Details
Gene Symbol: PLCXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005674904.3 | XP_005674961.2 | 3 - 4 | 1430 | 62 | |
GenBank mRNA | KA320718.1 | - | 3 - 4 | 1402 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
