Assay Details
Gene Symbol: PIGZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018046347.1 | XP_017901836.1 | 8 - 9 | 1706 | 63 | |
XM_018046352.1 | XP_017901841.1 | 7 - 8 | 1575 | 63 | ||
XM_018046357.1 | XP_017901846.1 | 7 - 8 | 1520 | 63 | ||
XM_018046364.1 | XP_017901853.1 | 6 - 7 | 1427 | 63 | ||
XM_018046370.1 | XP_017901859.1 | 7 - 8 | 1613 | 63 | ||
XM_018046373.1 | XP_017901862.1 | 6 - 7 | 1491 | 63 | ||
XM_018046376.1 | XP_017901865.1 | 8 - 9 | 1703 | 63 | ||
XM_018046379.1 | XP_017901868.1 | 7 - 8 | 1427 | 63 | ||
XM_018046382.1 | XP_017901871.1 | 5 - 6 | 1212 | 63 | ||
GenBank mRNA | JO379484.2 | - | 5 - 6 | 1086 | 63 | |
KA322544.1 | - | 1 - 2 | 2945 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
