Assay Details
Gene Symbol: FAM131A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018047187.1 | XP_017902676.1 | 4 - 5 | 1339 | 60 | |
XM_018047193.1 | XP_017902682.1 | 3 - 4 | 931 | 60 | ||
XM_018047196.1 | XP_017902685.1 | 4 - 5 | 992 | 60 | ||
XM_018047202.1 | XP_017902691.1 | 2 - 3 | 788 | 60 | ||
GenBank mRNA | JR114195.1 | - | 5384 | 60 | ||
JT706050.1 | - | 8 - 9 | 2442 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
