Assay Details
| Gene Symbol: METTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675713.2 | XP_005675770.2 | 5 - 6 | 954 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

METTL6| Gene Symbol: METTL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675713.2 | XP_005675770.2 | 5 - 6 | 954 | 72 | |
Set Membership: |
|