Assay Details
Gene Symbol: ANKMY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678993.3 | XP_005679050.1 | 1 - 2 | 524 | 65 | |
XM_018047354.1 | XP_017902843.1 | 1 - 2 | 526 | 65 | ||
GenBank mRNA | JT665247.1 | - | 11 - 12 | 2165 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
