Assay Details
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005679876.3 | XP_005679933.1 | 8 - 9 | 1593 | 57 | |
XM_005679877.3 | XP_005679934.1 | 8 - 9 | 1018 | 57 | ||
XM_018047677.1 | XP_017903166.1 | 8 - 9 | 1588 | 57 | ||
GenBank mRNA | JO647662.2 | - | 1 - 2 | 432 | 57 | |
JO647663.2 | - | 1 - 2 | 432 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
