Assay Details
Gene Symbol: ALG10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680680.3 | XP_005680737.1 | 2 - 3 | 669 | 89 | |
XM_013964120.2 | XP_013819574.1 | 2 - 3 | 318 | 89 | ||
GenBank mRNA | JO610019.2 | - | 2 - 3 | 505 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
