Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005681847.3 | XP_005681904.1 | 2 - 3 | 255 | 68 | |
| GenBank mRNA | JO580108.1 | - | 3 - 4 | 601 | 68 | |
More Information
Set Membership: |
|

PLAC8
Genomic Map