Assay Details
| Gene Symbol: POLR3G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005683428.3 | XP_005683485.1 | 2 - 3 | 283 | 103 | |
| XM_005683430.3 | XP_005683487.1 | 2 - 3 | 283 | 103 | ||
| XM_005683431.3 | XP_005683488.1 | 2 - 3 | 283 | 103 | ||
| XM_018050046.1 | XP_017905535.1 | 2 - 3 | 283 | 103 | ||
| GenBank mRNA | JT648920.1 | - | 5 - 6 | 961 | 103 | |
More Information
Set Membership: |
|

POLR3G
Genomic Map