Assay Details
Gene Symbol: SHF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005685868.3 | XP_005685925.3 | 2 - 3 | 660 | 62 | |
XM_013967166.2 | XP_013822620.2 | 2 - 3 | 660 | 62 | ||
XM_018054147.1 | XP_017909636.1 | 2 - 3 | 660 | 62 | ||
XM_018054148.1 | XP_017909637.1 | 2 - 3 | 660 | 62 | ||
XM_018054149.1 | XP_017909638.1 | 2 - 3 | 660 | 62 | ||
GenBank mRNA | KA349116.1 | - | 2 - 3 | 442 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
