Assay Details
| Gene Symbol: CHRAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688820.3 | XP_005688877.2 | 1 - 2 | 291 | 91 | |
| GenBank mRNA | JO578170.1 | - | 1 - 2 | 250 | 91 | |
| JT699346.1 | - | 3 - 4 | 693 | 91 | ||
More Information
Set Membership: |
|

CHRAC1
Genomic Map