Assay Details
Gene Symbol: PPP1R15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060015.1 | XP_017915504.1 | 1 - 2 | 2329 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: PPP1R15B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060015.1 | XP_017915504.1 | 1 - 2 | 2329 | 113 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |