Assay Details
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063836.1 | XP_017919325.1 | 2 - 3 | 405 | 92 | |
GenBank mRNA | KA313652.1 | - | 3 - 4 | 508 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
