Assay Details
| Gene Symbol: LOC102178324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697968.3 | XP_005698025.2 | 7 - 8 | 851 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102178324| Gene Symbol: LOC102178324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005697968.3 | XP_005698025.2 | 7 - 8 | 851 | 114 | |
Set Membership: |
|