Assay Details
Gene Symbol: LOC102172789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963527.2 | XP_013818981.2 | 5 - 6 | 467 | 88 | |
XM_018046879.1 | XP_017902368.1 | 4 - 5 | 783 | 88 | ||
XM_018046880.1 | XP_017902369.1 | 4 - 5 | 1389 | 88 | ||
XM_018046881.1 | XP_017902370.1 | 5 - 6 | 892 | 88 | ||
XM_018046882.1 | XP_017902371.1 | 5 - 6 | 481 | 88 | ||
XM_018046883.1 | XP_017902372.1 | 4 - 5 | 330 | 88 | ||
GenBank mRNA | JO643760.2 | - | 5 - 6 | 596 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
