Assay Details
Gene Symbol: LOC102172789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963527.2 | XP_013818981.2 | 7 - 8 | 773 | 163 | |
XM_018046879.1 | XP_017902368.1 | 6 - 7 | 1089 | 163 | ||
XM_018046880.1 | XP_017902369.1 | 6 - 7 | 1695 | 163 | ||
XM_018046881.1 | XP_017902370.1 | 7 - 8 | 1198 | 163 | ||
XM_018046882.1 | XP_017902371.1 | 7 - 8 | 787 | 163 | ||
XM_018046883.1 | XP_017902372.1 | 6 - 7 | 636 | 163 | ||
GenBank mRNA | JO643760.2 | - | 7 - 8 | 902 | 163 |

More Information
Set Membership: |
![]() ![]() ![]() |
