Assay Details
Gene Symbol: LOC102172789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963527.2 | XP_013818981.2 | 9 - 10 | 909 | 62 | |
XM_018046879.1 | XP_017902368.1 | 8 - 9 | 1225 | 62 | ||
XM_018046880.1 | XP_017902369.1 | 8 - 9 | 1831 | 62 | ||
XM_018046881.1 | XP_017902370.1 | 9 - 10 | 1334 | 62 | ||
XM_018046882.1 | XP_017902371.1 | 9 - 10 | 923 | 62 | ||
XM_018046883.1 | XP_017902372.1 | 8 - 9 | 772 | 62 | ||
GenBank mRNA | JO643760.2 | - | 9 - 10 | 1038 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
