Assay Details
Gene Symbol: LOC102172789 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963527.2 | XP_013818981.2 | 10 - 11 | 1092 | 85 | |
XM_018046879.1 | XP_017902368.1 | 9 - 10 | 1408 | 85 | ||
XM_018046880.1 | XP_017902369.1 | 9 - 10 | 2014 | 85 | ||
XM_018046881.1 | XP_017902370.1 | 10 - 11 | 1517 | 85 | ||
XM_018046882.1 | XP_017902371.1 | 10 - 11 | 1106 | 85 | ||
XM_018046883.1 | XP_017902372.1 | 9 - 10 | 955 | 85 | ||
GenBank mRNA | JO613548.2 | - | 1 - 2 | 1293 | 85 | |
JO643760.2 | - | 10 - 11 | 1221 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
