Assay Details
| Gene Symbol: C7H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051347.1 | XP_017906836.1 | 4 - 5 | 1234 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C7H19orf44| Gene Symbol: C7H19orf44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018051347.1 | XP_017906836.1 | 4 - 5 | 1234 | 76 | |
Set Membership: |
|