Assay Details
Gene Symbol: CDC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013965507.2 | XP_013820961.1 | 2 - 3 | 461 | 62 | |
GenBank mRNA | JO419781.2 | - | 2 - 3 | 413 | 62 | |
JO431658.2 | - | 3 - 4 | 1480 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
