Assay Details
Gene Symbol: HINT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052155.1 | XP_017907644.1 | 4 - 5 | 694 | 84 | |
XM_018052156.1 | XP_017907645.1 | 4 - 5 | 495 | 84 | ||
GenBank mRNA | JO437250.2 | - | 4 - 5 | 713 | 84 | |
JO535643.2 | - | 4 - 5 | 437 | 84 | ||
JO592721.2 | - | 1 - 2 | 543 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
