Assay Details
Gene Symbol: LAMTOR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018059411.1 | XP_017914900.1 | 2 - 3 | 349 | 51 | |
GenBank mRNA | JO425085.1 | - | 384 | 51 | ||
KA308179.1 | - | 3 - 4 | 846 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
