Assay Details
| Gene Symbol: SOX13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060001.1 | XP_017915490.1 | 8 - 9 | 931 | 68 | |
| XM_018060002.1 | XP_017915491.1 | 8 - 9 | 928 | 68 | ||
More Information
Set Membership: |
|

SOX13
Genomic Map