Assay Details
| Gene Symbol: DERL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018060909.1 | XP_017916398.1 | 4 - 5 | 412 | 85 | |
| XM_018060910.1 | XP_017916399.1 | 4 - 5 | 375 | 85 | ||
| XM_018060911.1 | XP_017916400.1 | 4 - 5 | 373 | 85 | ||
| GenBank mRNA | JO581226.1 | - | 4 - 5 | 459 | 85 | |
More Information
Set Membership: |
|

DERL3
Genomic Map