Assay Details
| Gene Symbol: CDYL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018061754.1 | XP_017917243.1 | 3 - 4 | 1073 | 59 | |
| GenBank mRNA | JO592159.2 | - | 4 - 5 | 1291 | 59 | |
| JT702018.1 | - | 5 - 6 | 1203 | 59 | ||
More Information
Set Membership: |
|

CDYL2
Genomic Map