Assay Details
| Gene Symbol: LOC102171917 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018065326.1 | XP_017920815.1 | 21 - 22 | 3283 | 101 | |
| XM_018065327.1 | XP_017920816.1 | 10 - 11 | 1917 | 101 | ||
More Information
Set Membership: |
|

LOC102171917
Genomic Map