Assay Details
Gene Symbol: RAB40B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065392.1 | XP_017920881.1 | 4 - 5 | 545 | 69 | |
XM_018065393.1 | XP_017920882.1 | 4 - 5 | 720 | 69 | ||
GenBank mRNA | JO436634.2 | - | 2 - 3 | 771 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
