Assay Details
Gene Symbol: ATPSCKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972873.2 | XP_013828327.2 | 4 - 5 | 650 | 86 | |
XM_018065549.1 | XP_017921038.1 | 4 - 5 | 583 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
