Assay Details
Gene Symbol: ARPIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066191.1 | XP_017921680.1 | 3 - 4 | 432 | 66 | |
XM_018066192.1 | XP_017921681.1 | 3 - 4 | 307 | 66 | ||
GenBank mRNA | JO433141.2 | - | 3 - 4 | 427 | 66 | |
JT673108.1 | - | 5 - 6 | 1091 | 66 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_018066192.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
