Assay Details
| Gene Symbol: PGBD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013974051.2 | XP_013829505.2 | 2 - 3 | 3164 | 100 | |
| XM_013974052.2 | XP_013829506.2 | 3 - 4 | 1473 | 100 | ||
| XM_013974054.2 | XP_013829508.2 | 3 - 4 | 1287 | 100 | ||
More Information
Set Membership: |
|

PGBD1
Genomic Map