Assay Details
Gene Symbol: APCDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018039497.1 | XP_017894986.1 | 2 - 3 | 439 | 80 | |
GenBank mRNA | JO444119.2 | - | 2 - 3 | 502 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
