Assay Details
| Gene Symbol: HEPACAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043577.1 | XP_017899066.1 | 3 - 4 | 1086 | 63 | |
| XM_018043578.1 | XP_017899067.1 | 3 - 4 | 1086 | 63 | ||
| GenBank mRNA | JO632295.2 | - | 3 - 4 | 1118 | 63 | |
More Information
Set Membership: |
|

HEPACAM
Genomic Map