Assay Details
| Gene Symbol: MRPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018043526.1 | XP_017899015.1 | 2 - 3 | 212 | 72 | |
| GenBank mRNA | JO528756.2 | - | 3 - 4 | 361 | 72 | |
| KA325906.1 | - | 3 - 4 | 525 | 72 | ||
More Information
Set Membership: |
|

MRPL23
Genomic Map