Assay Details
| Gene Symbol: FGF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005675114.3 | XP_005675171.1 | 2 - 3 | 1155 | 63 | |
| XM_005675116.3 | XP_005675173.1 | 3 - 4 | 364 | 63 | ||
| XM_013963800.2 | XP_013819254.2 | 2 - 3 | 280 | 63 | ||
| GenBank mRNA | JO645448.2 | - | 374 | 63 | ||
| JO645450.2 | - | 3 - 4 | 374 | 63 | ||
More Information
Set Membership: |
|

FGF12
Genomic Map