Assay Details
| Gene Symbol: S100A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005677503.3 | XP_005677560.2 | 2 - 3 | 240 | 90 | |
| XM_005677504.3 | XP_005677561.2 | 2 - 3 | 270 | 90 | ||
| XM_005677505.3 | XP_005677562.2 | 2 - 3 | 238 | 90 | ||
| XM_005677506.3 | XP_005677563.2 | 2 - 3 | 267 | 90 | ||
| XM_005677507.3 | XP_005677564.2 | 2 - 3 | 305 | 90 | ||
| XM_013976439.2 | XP_013831893.2 | 2 - 3 | 234 | 90 | ||
| XM_018046136.1 | XP_017901625.1 | 2 - 3 | 176 | 90 | ||
| GenBank mRNA | JT657902.1 | - | 2 - 3 | 121 | 90 | |
More Information
Set Membership: |
|

S100A5
Genomic Map