Assay Details
Gene Symbol: TM6SF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682084.3 | XP_005682141.2 | 6 - 7 | 854 | 71 | |
XM_005682085.3 | XP_005682142.2 | 6 - 7 | 854 | 71 | ||
XM_018051487.1 | XP_017906976.1 | 7 - 8 | 810 | 71 | ||
XM_018051488.1 | XP_017906977.1 | 7 - 8 | 810 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
