Assay Details
| Gene Symbol: REM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054571.1 | XP_017910060.1 | 1 - 2 | 1956 | 103 | |
| XM_018054572.1 | XP_017910061.1 | 2 - 3 | 3926 | 103 | ||
| XM_018054573.1 | XP_017910062.1 | 2 - 3 | 1740 | 103 | ||
| XM_018054574.1 | XP_017910063.1 | 3 - 4 | 3710 | 103 | ||
More Information
Set Membership: |
|

REM2
Genomic Map