Assay Details
| Gene Symbol: LOC102169972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687333.3 | XP_005687390.3 | 2 - 3 | 1130 | 88 | |
| XM_018056143.1 | XP_017911632.1 | 3 - 4 | 398 | 88 | ||
| XM_018056144.1 | XP_017911633.1 | 2 - 3 | 334 | 88 | ||
| GenBank mRNA | JO347079.2 | - | 5 - 6 | 718 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005687333.3, XM_018056143.1, XM_018056144.1 transcripts.
Set Membership: |
|

LOC102169972
Genomic Map