Assay Details
| Gene Symbol: LOC102169972 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005687333.3 | XP_005687390.3 | 6 - 7 | 1637 | 65 | |
| XM_018056143.1 | XP_017911632.1 | 7 - 8 | 905 | 65 | ||
| XM_018056144.1 | XP_017911633.1 | 6 - 7 | 841 | 65 | ||
| GenBank mRNA | JO347079.2 | - | 1 - 2 | 211 | 65 | |
More Information
Set Membership: |
|

LOC102169972
Genomic Map