Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005688619.3 | XP_005688676.2 | 5 - 6 | 594 | 74 | |
| XM_005688620.3 | XP_005688677.2 | 5 - 6 | 567 | 74 | ||
| XM_005688622.3 | XP_005688679.1 | 4 - 5 | 468 | 74 | ||
| XM_013968970.2 | XP_013824424.2 | 5 - 6 | 594 | 74 | ||
| GenBank mRNA | JO612051.2 | - | 5 - 6 | 560 | 74 | |
| KA313176.1 | - | 4 - 5 | 463 | 74 | ||
More Information
Set Membership: |
|

CD40
Genomic Map