Assay Details
Gene Symbol: NPM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698342.3 | XP_005698399.1 | 1 - 2 | 245 | 57 | |
GenBank mRNA | KA305381.1 | - | 5 - 6 | 756 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
