Assay Details
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005701096.3 | XP_005701153.1 | 3 - 4 | 630 | 100 | |
GenBank mRNA | JO439369.2 | - | 3 - 4 | 621 | 100 | |
JO641149.2 | - | 3 - 4 | 626 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
