Assay Details
| Gene Symbol: PLEKHD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018054039.1 | XP_017909528.1 | 4 - 5 | 686 | 74 | |
| GenBank mRNA | JO426243.1 | - | 4 - 5 | 600 | 74 | |
| JO432985.2 | - | 4 - 5 | 540 | 74 | ||
More Information
Set Membership: |
|

PLEKHD1
Genomic Map