Assay Details
Gene Symbol: PLA2G2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013972419.2 | XP_013827873.2 | 4 - 5 | 545 | 67 | |
XM_018054978.1 | XP_017910467.1 | 4 - 5 | 545 | 67 | ||
GenBank mRNA | KA337079.1 | - | 4 - 5 | 543 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
