Assay Details
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063651.1 | XP_017919140.1 | 4 - 5 | 347 | 59 | |
GenBank mRNA | KA330489.1 | - | 4 - 5 | 278 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
