Assay Details
Gene Symbol: LINGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018066520.1 | XP_017922009.1 | 4 | 1351 | 65 | |
XM_018066521.1 | XP_017922010.1 | 2 | 1326 | 65 | ||
XM_018066522.1 | XP_017922011.1 | 3 | 1306 | 65 | ||
XM_018066523.1 | XP_017922012.1 | 2 | 1141 | 65 | ||
XM_018066524.1 | XP_017922013.1 | 4 | 1352 | 65 | ||
GenBank mRNA | JO550599.2 | - | 2 | 1153 | 65 | |
JO633361.2 | - | 6 | 1717 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
