Assay Details
| Gene Symbol: ATRIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_018067022.1 | XP_017922511.1 | 6 - 7 | 1020 | 72 | |
| XM_018067023.1 | XP_017922512.1 | 6 - 7 | 1020 | 72 | ||
| XM_018067024.1 | XP_017922513.1 | 6 - 7 | 1020 | 72 | ||
More Information
Set Membership: |
|

ATRIP
Genomic Map